Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-20
Entrez Gene ID
188420
Gene Name
Zinc metalloproteinase nas-20
Gene Aliases
CELE_T11F9.3
Chromosome Location
Chr.V: 11470726 - 11473789 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073505.8 NP_505906.2 7 - 8 1422 118

View Full Product Details