Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- bath-39
- Entrez Gene ID
-
179827
- Gene Name
- BTB and MATH domain-containing protein 39
- Gene Aliases
-
CELE_F25H9.4
- Chromosome Location
-
Chr.V: 13459459 - 13461803 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073938.5 |
NP_506339.2 |
3 - 4 |
993 |
130 |
- Gene Symbol
- F25H9.2
- Entrez Gene ID
-
179826
- Gene Name
- BTB domain-containing protein
- Gene Aliases
-
CELE_F25H9.2
- Chromosome Location
-
Chr.V: 13458083 - 13461798 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073937.6 |
NP_506338.1 |
7 - 8 |
2177 |
130 |
View Full Product Details