Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tag-65
Entrez Gene ID
179857
Gene Name
Uncharacterized protein
Gene Aliases
CELE_F58G11.5
Chromosome Location
Chr.V: 13675988 - 13678587 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073985.6 NP_506386.1 3 - 4 386 70
GenBank mRNA AF536544.1 - 3 - 4 386 70

View Full Product Details