Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dhs-22
Entrez Gene ID
179940
Gene Name
DeHydrogenases, Short chain
Gene Aliases
CELE_C15H11.4
Chromosome Location
Chr.V: 14420052 - 14421440 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074169.7 NP_506570.1 2 - 3 224 105

View Full Product Details