Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scrm-7
Entrez Gene ID
190110
Gene Name
protein-coding
Gene Aliases
CELE_Y50E8A.9
Chromosome Location
Chr.V: 14778203 - 14779744 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276962.2 NP_001263891.1 3 - 4 725 121
NM_074245.3 NP_506646.1 2 - 3 536 121

View Full Product Details