Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-30
Entrez Gene ID
187955
Gene Name
UDP-glucuronosyltransferase
Gene Aliases
CELE_T01G5.2
Chromosome Location
Chr.V: 15121724 - 15125262 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074328.2 NP_506729.1 3 - 4 726 150

View Full Product Details