Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-45
Entrez Gene ID
188343
Gene Name
Seven TM Receptor
Gene Aliases
CELE_T09F5.3
Chromosome Location
Chr.V: 15158991 - 15160546 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074341.2 NP_506742.2 1 - 2 490 105

View Full Product Details