Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-21
Entrez Gene ID
183529
Gene Name
Serpentine receptor class epsilon-21
Gene Aliases
CELE_C47A10.4
Chromosome Location
Chr.V: 17769653 - 17773208 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075089.2 NP_507490.2 2 - 3 211 164

View Full Product Details