Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
exos-7
Entrez Gene ID
180369
Gene Name
RNase_PH domain-containing protein
Gene Aliases
CELE_F31D4.1
Chromosome Location
Chr.V: 20838073 - 20841622 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383513.1 4 - 5 831 137

View Full Product Details