Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
stau-1
Entrez Gene ID
180455
Gene Name
STAUfen (dsRNA binding protein) homolog
Gene Aliases
CELE_F55A4.5
Chromosome Location
Chr.X: 1027668 - 1035286 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075795.5 NP_508196.1 6 - 7 1199 100

View Full Product Details