Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
elc-2
Entrez Gene ID
24104987
Gene Name
Skp1_POZ domain-containing protein
Gene Aliases
CELE_W03H1.2
Chromosome Location
Chr.X: 1777288 - 1778088 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001374969.2 NP_001362016.1 1 - 2 158 80

View Full Product Details