Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nmy-1
Entrez Gene ID
180581
Gene Name
protein-coding
Gene Aliases
CELE_F52B10.1
Chromosome Location
Chr.X: 2909527 - 2918044 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076103.6 NP_508504.2 4 - 5 660 87

View Full Product Details