Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dhs-26
Entrez Gene ID
180624
Gene Name
DeHydrogenases, Short chain
Gene Aliases
CELE_ZK816.5
Chromosome Location
Chr.X: 3360722 - 3362493 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076179.5 NP_508580.2 5 - 6 974 119

View Full Product Details