Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glt-1
Entrez Gene ID
180641
Gene Name
protein-coding
Gene Aliases
CELE_C12D12.2
Chromosome Location
Chr.X: 3496026 - 3499169 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029222.4 NP_001024393.1 3 - 4 380 75
NM_001029223.6 NP_001024394.1 2 - 3 283 75
GenBank mRNA D86740.1 - - 402 75
D86741.1 - - 402 75

View Full Product Details