Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ncr-1
Entrez Gene ID
180719
Gene Name
NPC intracellular cholesterol transporter 1 homolog 1
Gene Aliases
CELE_F02E8.6
Chromosome Location
Chr.X: 4472244 - 4488857 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076370.5 NP_508771.1 3 - 4 480 66

View Full Product Details