Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ncr-1
- Entrez Gene ID
-
180719
- Gene Name
- NPC intracellular cholesterol transporter 1 homolog 1
- Gene Aliases
-
CELE_F02E8.6
- Chromosome Location
-
Chr.X: 4472244 - 4488857 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076370.5 |
NP_508771.1 |
5 - 6 |
922 |
78 |
View Full Product Details