Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-35
Entrez Gene ID
180725
Gene Name
protein-coding
Gene Aliases
CELE_C07A12.3
Chromosome Location
Chr.X: 4532298 - 4537546 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029194.4 NP_001024365.1 2 - 3 372 137
NM_001380949.1 NP_001367445.1 3 - 4 297 137

View Full Product Details