Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sulp-2
Entrez Gene ID
180828
Gene Name
STAS domain-containing protein
Gene Aliases
CELE_F14D12.5
Chromosome Location
Chr.X: 5595730 - 5599330 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076543.6 NP_508944.1 7 - 8 719 144
GenBank mRNA AY887908.1 - 7 - 8 741 144

View Full Product Details