Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zig-4
Entrez Gene ID
181051
Gene Name
Zwei Ig domain protein zig-4
Gene Aliases
CELE_C09C7.1
Chromosome Location
Chr.X: 7925434 - 7926595 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076934.7 NP_509335.1 2 - 3 478 73
GenBank mRNA AF456251.1 - 2 - 3 443 73

View Full Product Details