Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
inx-5
Entrez Gene ID
181086
Gene Name
Innexin-5
Gene Aliases
CELE_R09F10.4
Chromosome Location
Chr.X: 8300027 - 8302399 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077002.5 NP_509403.2 2 - 3 328 125

View Full Product Details