Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pgp-3
Entrez Gene ID
181326
Gene Name
Multidrug resistance protein pgp-3
Gene Aliases
CELE_ZK455.7
Chromosome Location
Chr.X: 11352156 - 11357094 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077500.5 NP_509901.1 4 - 5 774 130

View Full Product Details