Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-41
Entrez Gene ID
184602
Gene Name
Serpentine receptor class delta-41
Gene Aliases
CELE_F17A2.6
Chromosome Location
Chr.X: 12326724 - 12328365 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077662.1 NP_510063.1 1 - 2 304 113

View Full Product Details