Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-44
Entrez Gene ID
184604
Gene Name
Serpentine receptor class delta-44
Gene Aliases
CELE_F17A2.8
Chromosome Location
Chr.X: 12338440 - 12340248 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077667.1 NP_510068.1 2 - 3 421 112

View Full Product Details