Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
irk-3
Entrez Gene ID
191696
Gene Name
protein-coding
Gene Aliases
CELE_K04G11.5
Chromosome Location
Chr.X: 14353290 - 14370414 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001330918.4 NP_001317893.1 9 - 10 985 147
NM_001330920.4 NP_001317903.1 8 - 9 955 147
NM_001330921.5 NP_001317904.1 4 - 5 583 147
NM_001330922.4 NP_001317905.1 2 - 3 232 147
NM_001381146.1 NP_001368607.1 9 - 10 1000 147
NM_001381147.2 NP_001366850.1 9 - 10 1044 147

View Full Product Details