Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ccch-2
Entrez Gene ID
178454
Gene Name
CCCH-type zinc finger putative transcription factor
Gene Aliases
CELE_F38C2.5
Chromosome Location
Chr.IV: 16271255 - 16279010 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380549.2 NP_001366907.1 5 - 6 2601 128

View Full Product Details