Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ubxn-5
Entrez Gene ID
3565635
Gene Name
UBX domain-containing protein 5
Gene Aliases
CELE_T01E8.9
Chromosome Location
Chr.II: 10233833 - 10234498 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027129.4 NP_001022300.1 1 - 2 95 67

View Full Product Details