Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ave-1
Entrez Gene ID
3565044
Gene Name
SAM domain-containing protein
Gene Aliases
CELE_M01G5.6
Chromosome Location
Chr.III: 1495434 - 1495975 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027520.4 NP_001022691.1 1 - 2 200 98

View Full Product Details