Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-42
Entrez Gene ID
4363055
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_Y54F10BL.1
Chromosome Location
Chr.III: 2259906 - 2262527 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047421.4 NP_001040886.2 3 - 4 1898 118

View Full Product Details