Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GFI1
Entrez Gene ID
490156
Gene Name
growth factor independent 1 transcriptional repressor
Gene Aliases
-
Chromosome Location
Chr.6: 56473032 - 56484422 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012719.1 NP_001012737.1 5 - 6 1052 71
XM_022419500.1 XP_022275208.1 5 - 6 1113 71
GenBank mRNA AB178767.1 - 5 - 6 1082 71

View Full Product Details