Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DYNLT3
Entrez Gene ID
403505
Gene Name
dynein light chain Tctex-type 3
Gene Aliases
TCTE1L
Chromosome Location
Chr.X: 32603881 - 32611213 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003001.1 NP_001003001.1 3 - 4 206 118
XM_022415467.1 XP_022271175.1 3 - 4 273 118
GenBank mRNA AF491301.1 - 3 - 4 206 118

View Full Product Details