Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM53C
Entrez Gene ID
608692
Gene Name
family with sequence similarity 53 member C
Gene Aliases
-
Chromosome Location
Chr.11: 25940360 - 25951314 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_846104.5 XP_851197.1 2 - 3 381 73
XM_858230.5 XP_863323.1 2 - 3 371 73

View Full Product Details