Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS37B
Entrez Gene ID
477457
Gene Name
VPS37B subunit of ESCRT-I
Gene Aliases
-
Chromosome Location
Chr.26: 6594981 - 6621902 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005636144.3 XP_005636201.1 - 346 104
XM_014107872.2 XP_013963347.1 - 484 104
XM_534655.6 XP_534655.2 - 413 104

View Full Product Details