Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPS26
Entrez Gene ID
477175
Gene Name
mitochondrial ribosomal protein S26
Gene Aliases
-
Chromosome Location
Chr.24: 18215055 - 18216993 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_534365.6 XP_534365.2 3 - 4 517 75

View Full Product Details