Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLA2G12A
Entrez Gene ID
478514
Gene Name
phospholipase A2 group XIIA
Gene Aliases
-
Chromosome Location
Chr.32: 29913223 - 29929802 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_535693.6 XP_535693.1 3 - 4 580 99

View Full Product Details