Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPAT3
Entrez Gene ID
478460
Gene Name
glycerol-3-phosphate acyltransferase 3
Gene Aliases
AGPAT9
Chromosome Location
Chr.32: 7355154 - 7414921 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_535637.6 XP_535637.3 10 - 11 1155 77

View Full Product Details