Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATG12
Entrez Gene ID
474636
Gene Name
autophagy related 12
Gene Aliases
-
Chromosome Location
Chr.11: 5531062 - 5543034 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_531866.6 XP_531866.2 - 434 87

View Full Product Details