Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CBWD1
Entrez Gene ID
484176
Gene Name
COBW domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 89063511 - 89120995 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313837.1 NP_001300766.1 14 - 15 1165 76
XM_022421052.1 XP_022276760.1 15 - 16 1259 76
XM_022421090.1 XP_022276798.1 14 - 15 1214 76
XM_022421113.1 XP_022276821.1 15 - 16 1308 76
XM_022421139.1 XP_022276847.1 11 - 12 695 76

View Full Product Details