Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CC2D2B
Entrez Gene ID
486812
Gene Name
coiled-coil and C2 domain containing 2B
Gene Aliases
-
Chromosome Location
Chr.28: 9371454 - 9480331 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014108844.2 XP_013964319.1 23 - 24 2789 113
XM_014108856.2 XP_013964331.1 15 - 16 1763 113
XM_022411979.1 XP_022267687.1 28 - 29 3380 113
XM_022411980.1 XP_022267688.1 28 - 29 3381 113
XM_022411981.1 XP_022267689.1 28 - 29 3347 113
XM_022411982.1 XP_022267690.1 29 - 30 3435 113
XM_022411983.1 XP_022267691.1 27 - 28 3281 113
XM_022411985.1 XP_022267693.1 27 - 28 3266 113
XM_022411986.1 XP_022267694.1 27 - 28 3251 113
XM_022411987.1 XP_022267695.1 26 - 27 3238 113
XM_022411988.1 XP_022267696.1 27 - 28 3182 113
XM_022411989.1 XP_022267697.1 27 - 28 3170 113
XM_022411990.1 XP_022267698.1 26 - 27 3152 113
XM_022411991.1 XP_022267699.1 27 - 28 3259 113
XM_022411992.1 XP_022267700.1 27 - 28 3236 113
XM_022411993.1 XP_022267701.1 26 - 27 3195 113
XM_022411994.1 XP_022267702.1 26 - 27 3068 113
XM_022411997.1 XP_022267705.1 23 - 24 2932 113
XM_022411998.1 XP_022267706.1 27 - 28 3291 113
XM_022411999.1 XP_022267707.1 24 - 25 2705 113
XM_022412000.1 XP_022267708.1 21 - 22 2707 113
XM_022412001.1 XP_022267709.1 22 - 23 2618 113
XM_022412002.1 XP_022267710.1 21 - 22 2564 113
XM_022412003.1 XP_022267711.1 20 - 21 2497 113
XM_022412006.1 XP_022267714.1 22 - 23 2482 113
XM_022412007.1 XP_022267715.1 22 - 23 2524 113
XM_022412008.1 XP_022267716.1 23 - 24 2569 113
XM_022412009.1 XP_022267717.1 23 - 24 2554 113
XM_022412010.1 XP_022267718.1 22 - 23 2508 113
XM_022412011.1 XP_022267719.1 22 - 23 2525 113
XM_022412012.1 XP_022267720.1 22 - 23 2486 113
XM_022412013.1 XP_022267721.1 22 - 23 2469 113
XM_022412014.1 XP_022267722.1 12 - 13 1401 113
XM_022412015.1 XP_022267723.1 13 - 14 1572 113

View Full Product Details