Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYBA1
Entrez Gene ID
491178
Gene Name
crystallin beta A1
Gene Aliases
CRYBA3
Chromosome Location
Chr.9: 43372507 - 43378561 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080899.1 NP_001074368.1 5 - 6 506 126
GenBank mRNA EF191198.1 - 5 - 6 506 126

View Full Product Details