Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRGN
Entrez Gene ID
609421
Gene Name
serglycin
Gene Aliases
-
Chromosome Location
Chr.4: 20230713 - 20244952 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_846674.5 XP_851767.1 2 - 3 302 106

View Full Product Details