Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB27A
Entrez Gene ID
608699
Gene Name
RAB27A, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.30: 20633576 - 20706240 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048130.1 NP_001041595.1 1 - 2 68 153
XM_005638180.3 XP_005638237.1 2 - 3 224 153
XM_005638181.2 XP_005638238.1 2 - 3 248 153
GenBank mRNA DQ494380.1 - 1 - 2 68 153

View Full Product Details