Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STK35
Entrez Gene ID
609432
Gene Name
serine/threonine kinase 35
Gene Aliases
-
Chromosome Location
Chr.24: 18913746 - 18958240 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_846689.5 XP_851782.4 2 - 3 924 93

View Full Product Details