Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AQP8
Entrez Gene ID
608036
Gene Name
aquaporin 8
Gene Aliases
-
Chromosome Location
Chr.6: 20984868 - 20992647 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005621395.3 XP_005621452.1 4 - 5 756 63
XM_845033.5 XP_850126.1 4 - 5 807 63

View Full Product Details