Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TEX28
Entrez Gene ID
492248
Gene Name
testis expressed 28
Gene Aliases
-
Chromosome Location
Chr.X: 121987726 - 122030718 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022415830.1 XP_022271538.1 3 - 3 547 151
XM_549368.5 XP_549368.3 3 - 3 873 151

View Full Product Details