Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CALM2
Entrez Gene ID
474584
Gene Name
calmodulin 2
Gene Aliases
CaMII, calmodulin
Chromosome Location
Chr.10: 49299698 - 49314533 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_859552.5 XP_864645.2 5 - 6 535 130

View Full Product Details