Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSHZ3
Entrez Gene ID
484607
Gene Name
teashirt zinc finger homeobox 3
Gene Aliases
-
Chromosome Location
Chr.1: 120388125 - 120471185 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005616783.3 XP_005616840.1 2 - 2 2972 145
XM_022426433.1 XP_022282141.1 3 - 3 3132 145

View Full Product Details