Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
USP49
Entrez Gene ID
474899
Gene Name
ubiquitin specific peptidase 49
Gene Aliases
-
Chromosome Location
Chr.12: 10474873 - 10556900 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022426033.1 XP_022281741.1 3 - 4 1500 114

View Full Product Details