Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CNPY4
Entrez Gene ID
489845
Gene Name
canopy FGF signaling regulator 4
Gene Aliases
-
Chromosome Location
Chr.6: 9475277 - 9479632 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_546963.5 XP_546963.2 - 471 122

View Full Product Details