Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZACN
Entrez Gene ID
483326
Gene Name
zinc activated ion channel
Gene Aliases
LGICZ, LGICZ1
Chromosome Location
Chr.9: 4555299 - 4558599 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010955.1 NP_001010955.1 - 267 67

View Full Product Details