Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hoxc9
Entrez Gene ID
100763984
Gene Name
homeobox C9
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614339: 67904 - 71762 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647750.1 XP_007645940.1 1 - 2 1181 69

View Full Product Details